Assay Details
| Gene Symbol: Plppr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866008.3 | XP_004866065.1 | 3 - 4 | 764 | 78 | |
| XM_021244851.1 | XP_021100510.1 | 3 - 4 | 524 | 78 | ||
More Information
Set Membership: |
|

Plppr3
Genomic Map