Assay Details
| Gene Symbol: Ifi30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251014.1 | XP_021106673.1 | 4 - 5 | 516 | 63 | |
| XM_021251015.1 | XP_021106674.1 | 4 - 5 | 497 | 63 | ||
More Information
Set Membership: |
|

Ifi30
Genomic Map