Assay Details
| Gene Symbol: CUNH12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021243084.1 | XP_021098743.1 | 4 - 5 | 128 | 82 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH12orf54| Gene Symbol: CUNH12orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021243084.1 | XP_021098743.1 | 4 - 5 | 128 | 82 | |