Assay Details
| Gene Symbol: Ccdc63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257486.1 | XP_021113145.1 | 10 - 11 | 3070 | 62 | |
| XM_021257488.1 | XP_021113147.1 | 10 - 11 | 1523 | 62 | ||
| XM_021257489.1 | XP_021113148.1 | 10 - 11 | 3073 | 62 | ||
| XM_021257490.1 | XP_021113149.1 | 9 - 10 | 1454 | 62 | ||
More Information
Set Membership: |
|

Ccdc63
Genomic Map