Assay Details
| Gene Symbol: Neurl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836376.3 | XP_004836433.1 | 4 - 5 | 1582 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Neurl1b| Gene Symbol: Neurl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836376.3 | XP_004836433.1 | 4 - 5 | 1582 | 118 | |
Set Membership: |
|