Assay Details
| Gene Symbol: Mrps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849011.3 | XP_004849068.1 | 3 - 4 | 344 | 66 | |
| XM_021261789.1 | XP_021117448.1 | 2 - 3 | 202 | 66 | ||
More Information
Set Membership: |
|

Mrps2
Genomic Map