Assay Details
| Gene Symbol: Slc13a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856986.3 | XP_004857043.1 | 6 - 7 | 924 | 64 | |
| XM_004856987.3 | XP_004857044.1 | 5 - 6 | 795 | 64 | ||
| XM_004856988.3 | XP_004857045.1 | 6 - 7 | 924 | 64 | ||
More Information
Set Membership: |
|

Slc13a2
Genomic Map