Assay Details
| Gene Symbol: Pcdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844268.2 | XP_004844325.1 | 2 - 3 | 3117 | 104 | |
| XM_013072069.2 | XP_012927523.1 | 2 - 3 | 2822 | 104 | ||
More Information
Set Membership: |
|

Pcdh8
Genomic Map