Assay Details
| Gene Symbol: LOC101704936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854114.2 | XP_004854171.1 | 1 - 2 | 97 | 68 | |
| XM_021265970.1 | XP_021121629.1 | 1 - 2 | 97 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004854114.2, XM_021265970.1 transcripts.
Set Membership: |
|

LOC101704936
Genomic Map