Assay Details
| Gene Symbol: LOC101704936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854114.2 | XP_004854171.1 | 2 - 3 | 245 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101704936| Gene Symbol: LOC101704936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854114.2 | XP_004854171.1 | 2 - 3 | 245 | 56 | |
Set Membership: |
|