Assay Details
| Gene Symbol: Tapbpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869389.2 | XP_004869446.1 | 4 - 5 | 1073 | 60 | |
| XM_021248126.1 | XP_021103785.1 | 4 - 5 | 1077 | 60 | ||
| XM_021248127.1 | XP_021103786.1 | 4 - 5 | 927 | 60 | ||
More Information
Set Membership: |
|

Tapbpl
Genomic Map