Assay Details
| Gene Symbol: Fam155b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871766.2 | XP_004871823.1 | 1 - 2 | 1834 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam155b| Gene Symbol: Fam155b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871766.2 | XP_004871823.1 | 1 - 2 | 1834 | 70 | |
Set Membership: |
|