Assay Details
| Gene Symbol: Gfy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066420.2 | XP_012921874.1 | 1 - 2 | 969 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013066420.2 transcripts.
Set Membership: |
|

Gfy
Genomic Map