Assay Details
| Gene Symbol: Sept14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021254296.1 | XP_021109955.1 | 8 - 9 | 975 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sept14| Gene Symbol: Sept14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021254296.1 | XP_021109955.1 | 8 - 9 | 975 | 73 | |
Set Membership: |
|