Assay Details
| Gene Symbol: Psmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850932.2 | XP_004850989.1 | 3 - 4 | 416 | 59 | |
| XM_004850934.3 | XP_004850991.1 | 3 - 4 | 367 | 59 | ||
More Information
Set Membership: |
|

Psmb2
Genomic Map