Assay Details
| Gene Symbol: Znf550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866575.2 | XP_004866632.1 | 1 - 2 | 701 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004866575.2 transcripts.
Set Membership: |
|

Znf550
Genomic Map