Assay Details
| Gene Symbol: Crip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836811.2 | XP_004836868.1 | 6 - 7 | 790 | 71 | |
| XM_004836813.3 | XP_004836870.1 | 5 - 6 | 526 | 71 | ||
| XM_004836815.2 | XP_004836872.1 | 5 - 6 | 482 | 71 | ||
| XM_004836818.3 | XP_004836875.1 | 6 - 7 | 536 | 71 | ||
| XM_004836819.3 | XP_004836876.1 | 6 - 7 | 534 | 71 | ||
| XM_004836820.2 | XP_004836877.1 | 7 - 8 | 726 | 71 | ||
| XM_013066066.1 | XP_012921520.1 | 6 - 7 | 793 | 71 | ||
| XM_021243297.1 | XP_021098956.1 | 7 - 8 | 798 | 71 | ||
More Information
Set Membership: |
|

Crip2
Genomic Map