Assay Details
| Gene Symbol: Tmem248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839999.3 | XP_004840056.1 | 6 - 7 | 1016 | 79 | |
| XM_021254337.1 | XP_021109996.1 | 6 - 7 | 1145 | 79 | ||
| XM_021254338.1 | XP_021109997.1 | 6 - 7 | 965 | 79 | ||
More Information
Set Membership: |
|

Tmem248
Genomic Map