Assay Details
| Gene Symbol: Hras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851722.3 | XP_004851779.1 | 4 - 5 | 579 | 68 | |
| XM_021264062.1 | XP_021119721.1 | 4 - 5 | 517 | 68 | ||
| XM_021264063.1 | XP_021119722.1 | 4 - 5 | 517 | 68 | ||
More Information
Set Membership: |
|

Hras
Genomic Map