Assay Details
| Gene Symbol: Rgs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865175.2 | XP_004865232.1 | 1 - 2 | 199 | 63 | |
| XM_013065738.2 | XP_012921192.1 | 1 - 2 | 198 | 63 | ||
More Information
Set Membership: |
|

Rgs4
Genomic Map