Assay Details
| Gene Symbol: Ramp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868394.3 | XP_004868451.1 | 2 - 3 | 266 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ramp1| Gene Symbol: Ramp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868394.3 | XP_004868451.1 | 2 - 3 | 266 | 84 | |
Set Membership: |
|