Assay Details
| Gene Symbol: Klk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066412.2 | XP_012921866.1 | 5 - 6 | 843 | 73 | |
| XM_021246187.1 | XP_021101846.1 | 4 - 5 | 658 | 73 | ||
| XM_021246188.1 | XP_021101847.1 | 4 - 5 | 625 | 73 | ||
| XM_021246189.1 | XP_021101848.1 | 4 - 5 | 603 | 73 | ||
More Information
Set Membership: |
|

Klk8
Genomic Map