Assay Details
| Gene Symbol: Serinc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841599.3 | XP_004841656.1 | 3 - 4 | 460 | 51 | |
| XM_013071073.2 | XP_012926527.1 | 3 - 4 | 499 | 51 | ||
| XM_013071075.2 | XP_012926529.1 | 2 - 3 | 293 | 51 | ||
| XM_013071076.2 | XP_012926530.1 | 3 - 4 | 461 | 51 | ||
| XM_021255700.1 | XP_021111359.1 | 3 - 4 | 461 | 51 | ||
More Information
Set Membership: |
|

Serinc5
Genomic Map