Assay Details
| Gene Symbol: Anks1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846729.3 | XP_004846786.1 | 17 - 18 | 2811 | 58 | |
| XM_013072818.2 | XP_012928272.1 | 18 - 19 | 2909 | 58 | ||
| XM_013072819.2 | XP_012928273.1 | 17 - 18 | 2845 | 58 | ||
| XM_013072820.2 | XP_012928274.1 | 18 - 19 | 2908 | 58 | ||
| XM_021259962.1 | XP_021115621.1 | 8 - 9 | 1877 | 58 | ||
More Information
Set Membership: |
|

Anks1a
Genomic Map