Assay Details
Gene Symbol: Il17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004854391.2 | XP_004854448.1 | 3 - 4 | 631 | 114 | |
XM_021266195.1 | XP_021121854.1 | 3 - 4 | 485 | 114 | ||
XM_021266196.1 | XP_021121855.1 | 3 - 4 | 388 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
