Assay Details
| Gene Symbol: Atpaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872968.3 | XP_004873025.1 | 8 - 9 | 1040 | 69 | |
| XM_021250796.1 | XP_021106455.1 | 7 - 8 | 595 | 69 | ||
More Information
Set Membership: |
|

Atpaf1
Genomic Map