Assay Details
| Gene Symbol: Ahsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834830.3 | XP_004834887.1 | 2 - 3 | 420 | 58 | |
| XM_013072219.2 | XP_012927673.1 | 2 - 3 | 421 | 58 | ||
More Information
Set Membership: |
|

Ahsg
Genomic Map