Assay Details
| Gene Symbol: Bfsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840852.3 | XP_004840909.3 | 1 - 2 | 1002 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bfsp1| Gene Symbol: Bfsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840852.3 | XP_004840909.3 | 1 - 2 | 1002 | 56 | |
Set Membership: |
|