Assay Details
| Gene Symbol: Agrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843031.2 | XP_004843088.1 | 3 - 4 | 422 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agrp| Gene Symbol: Agrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843031.2 | XP_004843088.1 | 3 - 4 | 422 | 91 | |
Set Membership: |
|