Assay Details
| Gene Symbol: Chpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844853.3 | XP_004844910.1 | 5 - 6 | 822 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chpt1| Gene Symbol: Chpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844853.3 | XP_004844910.1 | 5 - 6 | 822 | 63 | |
Set Membership: |
|