Assay Details
| Gene Symbol: Igsf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858727.3 | XP_004858784.1 | 3 - 4 | 1005 | 57 | |
| XM_021239733.1 | XP_021095392.1 | 3 - 4 | 1005 | 57 | ||
| XM_021239734.1 | XP_021095393.1 | 4 - 5 | 879 | 57 | ||
More Information
Set Membership: |
|

Igsf8
Genomic Map