Assay Details
| Gene Symbol: Mindy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075530.2 | XP_012930984.1 | 2 - 3 | 1179 | 97 | |
| XM_013075531.2 | XP_012930985.1 | 3 - 4 | 1240 | 97 | ||
| XM_021265911.1 | XP_021121570.1 | 3 - 4 | 1240 | 97 | ||
| XM_021265912.1 | XP_021121571.1 | 4 - 5 | 1300 | 97 | ||
More Information
Set Membership: |
|

Mindy1
Genomic Map