Assay Details
| Gene Symbol: Mindy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075530.2 | XP_012930984.1 | 5 - 6 | 1466 | 98 | |
| XM_013075531.2 | XP_012930985.1 | 6 - 7 | 1527 | 98 | ||
| XM_021265911.1 | XP_021121570.1 | 6 - 7 | 1527 | 98 | ||
| XM_021265912.1 | XP_021121571.1 | 7 - 8 | 1587 | 98 | ||
More Information
Set Membership: |
|

Mindy1
Genomic Map