Assay Details
| Gene Symbol: Mindy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075530.2 | XP_012930984.1 | 7 - 8 | 1703 | 78 | |
| XM_013075531.2 | XP_012930985.1 | 8 - 9 | 1764 | 78 | ||
| XM_021265911.1 | XP_021121570.1 | 8 - 9 | 1764 | 78 | ||
| XM_021265912.1 | XP_021121571.1 | 9 - 10 | 1824 | 78 | ||
More Information
Set Membership: |
|

Mindy1
Genomic Map