Assay Details
| Gene Symbol: Rnf145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836473.2 | XP_004836530.1 | 11 - 12 | 2036 | 60 | |
| XM_021240425.1 | XP_021096084.1 | 10 - 11 | 1685 | 60 | ||
More Information
Set Membership: |
|

Rnf145
Genomic Map