Assay Details
| Gene Symbol: Sptlc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840827.3 | XP_004840884.2 | 10 - 11 | 1999 | 78 | |
| XM_021255072.1 | XP_021110731.1 | 11 - 12 | 1817 | 78 | ||
| XM_021255073.1 | XP_021110732.1 | 11 - 12 | 1811 | 78 | ||
| XM_021255074.1 | XP_021110733.1 | 10 - 11 | 1708 | 78 | ||
More Information
Set Membership: |
|

Sptlc3
Genomic Map