Assay Details
| Gene Symbol: Ubxn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842152.2 | XP_004842209.1 | 6 - 7 | 738 | 70 | |
| XM_021256096.1 | XP_021111755.1 | 6 - 7 | 738 | 70 | ||
More Information
Set Membership: |
|

Ubxn2b
Genomic Map