Assay Details
| Gene Symbol: Setbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855072.3 | XP_004855129.1 | 3 - 4 | 890 | 69 | |
| XM_021236586.1 | XP_021092245.1 | 3 - 4 | 891 | 69 | ||
More Information
Set Membership: |
|

Setbp1
Genomic Map