Assay Details
| Gene Symbol: Gjb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860243.3 | XP_004860300.1 | 2 - 3 | 895 | 85 | |
| XM_021240747.1 | XP_021096406.1 | 2 - 3 | 949 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004860243.3, XM_021240747.1 transcripts.
Set Membership: |
|

Gjb7
Genomic Map