Assay Details
| Gene Symbol: Ogfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863996.3 | XP_004864053.1 | 4 - 5 | 570 | 62 | |
| XM_004863997.3 | XP_004864054.1 | 4 - 5 | 567 | 62 | ||
| XM_021243753.1 | XP_021099412.1 | 5 - 6 | 702 | 62 | ||
| XM_021243755.1 | XP_021099414.1 | 5 - 6 | 699 | 62 | ||
More Information
Set Membership: |
|

Ogfrl1
Genomic Map