Assay Details
| Gene Symbol: Gyg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867322.2 | XP_004867379.1 | 4 - 5 | 576 | 62 | |
| XM_004867323.3 | XP_004867380.1 | 4 - 5 | 526 | 62 | ||
| XM_004867324.3 | XP_004867381.1 | 4 - 5 | 497 | 62 | ||
| XM_021246353.1 | XP_021102012.1 | 4 - 5 | 577 | 62 | ||
| XM_021246354.1 | XP_021102013.1 | 4 - 5 | 576 | 62 | ||
| XM_021246355.1 | XP_021102014.1 | 4 - 5 | 577 | 62 | ||
More Information
Set Membership: |
|

Gyg2
Genomic Map