Assay Details
| Gene Symbol: Otud6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841952.3 | XP_004842009.2 | 1 - 2 | 163 | 65 | |
| XM_021255953.1 | XP_021111612.1 | 1 - 2 | 163 | 65 | ||
| XM_021255954.1 | XP_021111613.1 | 1 - 2 | 183 | 65 | ||
More Information
Set Membership: |
|

Otud6b
Genomic Map