Assay Details
| Gene Symbol: Zdhhc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852296.2 | XP_004852353.1 | 2 - 3 | 598 | 65 | |
| XM_021264541.1 | XP_021120200.1 | 4 - 5 | 820 | 65 | ||
More Information
Set Membership: |
|

Zdhhc24
Genomic Map