Assay Details
| Gene Symbol: Znf106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861739.3 | XP_004861796.2 | 16 - 17 | 5349 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znf106| Gene Symbol: Znf106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861739.3 | XP_004861796.2 | 16 - 17 | 5349 | 61 | |
Set Membership: |
|