Assay Details
| Gene Symbol: Tmem106a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870587.3 | XP_004870644.1 | 3 - 4 | 484 | 72 | |
| XM_021248896.1 | XP_021104555.1 | 4 - 5 | 372 | 72 | ||
More Information
Set Membership: |
|

Tmem106a
Genomic Map