Assay Details
| Gene Symbol: LOC101710978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004875595.3 | XP_004875652.3 | 1 - 2 | 488 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004875595.3 transcripts.
Set Membership: |
|

LOC101710978
Genomic Map