Assay Details
| Gene Symbol: Slc36a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078831.2 | XP_012934285.1 | 4 - 5 | 507 | 55 | |
| XM_013078839.2 | XP_012934293.1 | 4 - 5 | 469 | 55 | ||
| XM_021240987.1 | XP_021096646.1 | 5 - 6 | 650 | 55 | ||
| XM_021241000.1 | XP_021096659.1 | 5 - 6 | 650 | 55 | ||
More Information
Set Membership: |
|

Slc36a1
Genomic Map