Assay Details
| Gene Symbol: Spred2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849957.3 | XP_004850014.1 | 5 - 6 | 1544 | 67 | |
| XM_004849958.3 | XP_004850015.1 | 5 - 6 | 704 | 67 | ||
More Information
Set Membership: |
|

Spred2
Genomic Map