Assay Details
| Gene Symbol: Adpgk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855519.3 | XP_004855576.2 | 2 - 3 | 531 | 80 | |
| XM_004855520.3 | XP_004855577.2 | 2 - 3 | 531 | 80 | ||
More Information
Set Membership: |
|

Adpgk
Genomic Map