Assay Details
| Gene Symbol: Mtpap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861786.2 | XP_004861843.1 | 1 - 2 | 211 | 83 | |
| XM_004861787.2 | XP_004861844.1 | 1 - 2 | 209 | 83 | ||
| XM_021242001.1 | XP_021097660.1 | 1 - 2 | 211 | 83 | ||
| XM_021242002.1 | XP_021097661.1 | 1 - 2 | 211 | 83 | ||
| XM_021242003.1 | XP_021097662.1 | 1 - 2 | 210 | 83 | ||
More Information
Set Membership: |
|

Mtpap
Genomic Map